AMBH580644F3 Ambeed, Inc.

(R)-2,5-Dioxotetrahydrofuran-3-yl acetate

  • MDL number MFCD00145214
  • Cas Number : 79814-40-7
  • Molecular Weight : 158.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/79814-40-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/79814-40-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart