AMBH580644F5 Ambeed, Inc.

(S)-Methyl 2-amino-2-cyclopentylacetate

  • MDL number MFCD20617284
  • Cas Number : 801162-35-6
  • Molecular Weight : 157.2102

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Physical Form White to Yellow Solid or liquid
Shipping Temperature Keep in dark place,Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/801162-35-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/801162-35-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart