AMBH580644F9 Ambeed, Inc.

Methyl 5-(chlorosulfonyl)-1-methyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD02180789
  • Cas Number : 306936-53-8
  • Molecular Weight : 237.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/306936-53-8.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/306936-53-8.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart