AMBH580644FF Ambeed, Inc.

Di(pyridin-4-yl)amine

  • MDL number MFCD00829434
  • Cas Number : 1915-42-0
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(4-pyridinyl)-4-pyridinamine
Inchi Code 1S/C10H9N3/c1-5-11-6-2-9(1)13-10-3-7-12-8-4-10/h1-8H,(H,11,12,13)
Inchi Key KDWIKPVYQSPYIX-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1915-42-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1915-42-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart