AMBH58064503 Ambeed, Inc.

Strontiumhydroxideoctahydrate

  • MDL number MFCD00150395
  • Cas Number : 1311-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/1311-10-0.html

Safety Information

MSDS https://www.ambeed.com/products/1311-10-0.html
Availability details for
Quantity
  • Availability
  • Add to Cart