AMBH5806450A Ambeed, Inc.

1-(2-Bromo-4-fluorophenyl)ethanone

  • MDL number MFCD00077464
  • Cas Number : 1006-39-9
  • Molecular Weight : 217.0351

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Physical Form Colorless to Pale-yellow to Yellow-brown Solid or Liquid
Shipping Temperature Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1006-39-9.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/1006-39-9.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart