AMBH5806450B Ambeed, Inc.

Calcium 2-hydroxypropanoate pentahydrate

  • MDL number MFCD00150987
  • Cas Number : 5743-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/5743-47-5.html

Safety Information

MSDS https://www.ambeed.com/products/5743-47-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart