AMBH5806450C Ambeed, Inc.

Leniolisib

  • MDL number MFCD30470232
  • Cas Number : 1354690-24-6
  • Molecular Weight : 450.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/1354690-24-6.html

Safety Information

MSDS https://www.ambeed.com/products/1354690-24-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart