AMBH58064517 Ambeed, Inc.

Pentadecane-1,15-diol

  • MDL number MFCD00039633
  • Cas Number : 14722-40-8
  • Molecular Weight : 244.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/14722-40-8.html

Safety Information

MSDS https://www.ambeed.com/products/14722-40-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart