AMBH58064518 Ambeed, Inc.

6-Bromo-1,8-naphthyridin-4(1H)-one

  • MDL number MFCD12024515
  • Cas Number : 1150618-43-1
  • Molecular Weight : 225.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1150618-43-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1150618-43-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart