AMBH5806451D Ambeed, Inc.

4-Chloroquinoline-3-carbaldehyde

  • MDL number MFCD03787717
  • Cas Number : 201420-30-6
  • Molecular Weight : 191.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/201420-30-6.html

Safety Information

MSDS https://www.ambeed.com/products/201420-30-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart