AMBH5806451E Ambeed, Inc.

5-Methoxybenzo[d]thiazole

  • MDL number MFCD06659660
  • Cas Number : 2942-14-5
  • Molecular Weight : 165.2123

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/2942-14-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/2942-14-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart