AMBH58064523 Ambeed, Inc.

4-Amino-2-(methylthio)thiazole-5-carbonitrile

  • MDL number MFCD00266086
  • Cas Number : 39736-28-2
  • Molecular Weight : 171.2433

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/39736-28-2.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/39736-28-2.html
pictograms GHS07
hstatements H302-H312-H332
Availability details for
Quantity
  • Availability
  • Add to Cart