AMBH58064526 Ambeed, Inc.

1-(Dimethylamino)-2-methylpropan-2-ol

  • MDL number MFCD11934288
  • Cas Number : 14123-48-9
  • Molecular Weight : 117.1900

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form liquid
Shipping Temperature Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/14123-48-9.html

Safety Information

signalword Warning
pcodes P261-P342+P311-P305+P351+P338
MSDS https://www.ambeed.com/products/14123-48-9.html
pictograms GHS07
hstatements H319-H315-H335
Availability details for
Quantity
  • Availability
  • Add to Cart