AMBH58064528 Ambeed, Inc.

6-Bromo-8-methylimidazo[1,2-a]pyridin-2-amine

  • MDL number MFCD26707796
  • Cas Number : 1500190-82-8
  • Molecular Weight : 226.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1500190-82-8.html

Safety Information

MSDS https://www.ambeed.com/products/1500190-82-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart