AMBH58064530 Ambeed, Inc.

Phenylbiguanide

  • MDL number MFCD00179077
  • Cas Number : 102-02-3
  • Molecular Weight : 177.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/102-02-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/102-02-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart