AMBH58064531 Ambeed, Inc.

7-Chloroisoquinolin-3-amine

  • MDL number MFCD15526594
  • Cas Number : 82117-29-1
  • Molecular Weight : 178.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 365.7�0�t 760 mmHg
Iupac Name 7-chloro-3-isoquinolinylamine
Inchi Code 1S/C9H7ClN2/c10-8-2-1-6-4-9(11)12-5-7(6)3-8/h1-5H,(H2,11,12)
Inchi Key BNICXCCUFXPCBF-UHFFFAOYSA-N

Description

Purity 97%
Flash Point 175
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 365.7�0�t 760 mmHg
Product Link https://www.ambeed.com/products/82117-29-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/82117-29-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart