AMBH58064533 Ambeed, Inc.

2-(tert-Butyl)-4-chloro-5-nitropyrimidine

  • MDL number MFCD18250574
  • Cas Number : 70227-50-8
  • Molecular Weight : 215.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/70227-50-8.html

Safety Information

MSDS https://www.ambeed.com/products/70227-50-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart