AMBH58064535 Ambeed, Inc.

(S)-2-((tert-Butoxycarbonyl)amino)-3-ethoxypropanoic acid

  • MDL number MFCD06797558
  • Cas Number : 104839-00-1
  • Molecular Weight : 233.2616

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/104839-00-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/104839-00-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart