AMBH5806453B Ambeed, Inc.

(E)-3-(3-(Trifluoromethyl)phenyl)acrylaldehyde

  • MDL number MFCD09260891
  • Cas Number : 262268-58-6
  • Molecular Weight : 200.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/262268-58-6.html

Safety Information

MSDS https://www.ambeed.com/products/262268-58-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart