AMBH5806453D Ambeed, Inc.

2-(Trifluoromethyl)benzo[d]thiazol-6-ol

  • Cas Number : 1261740-05-9
  • Molecular Weight : 219.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1261740-05-9.html

Safety Information

MSDS https://www.ambeed.com/products/1261740-05-9.html
Availability details for
Quantity
  • Availability
  • Add to Cart