AMBH58064546 Ambeed, Inc.

3-Nitrobenzenesulfonohydrazide

  • MDL number MFCD01859908
  • Cas Number : 6655-77-2
  • Molecular Weight : 217.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/6655-77-2.html

Safety Information

MSDS https://www.ambeed.com/products/6655-77-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart