AMBH58064552 Ambeed, Inc.

1-Allyl-1H-pyrrole-2,5-dione

  • MDL number MFCD00523374
  • Cas Number : 2973-17-3
  • Molecular Weight : 137.1360

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form Solid or liquid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/2973-17-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/2973-17-3.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart