AMBH58064558 Ambeed, Inc.

2-Methyl-6-oxo-1,6-dihydropyrimidine-4-carboxylic acid

  • MDL number MFCD01689713
  • Cas Number : 34415-10-6
  • Molecular Weight : 154.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Keep in dark place,Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 301.034°C at 760 mmHg
Product Link https://www.ambeed.com/products/34415-10-6.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/34415-10-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart