AMBH58064561 Ambeed, Inc.

3-Fluoroquinolin-6-ol

  • MDL number MFCD18414584
  • Cas Number : 808755-53-5
  • Molecular Weight : 163.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 316.6�0�t 760 mmHg
Iupac Name 3-fluoro-6-quinolinol
Inchi Code 1S/C9H6FNO/c10-7-3-6-4-8(12)1-2-9(6)11-5-7/h1-5,12H
Inchi Key WUAYWIYDQAWNFI-UHFFFAOYSA-N

Description

Purity 95%
Flash Point 145.3
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 316.6�0�t 760 mmHg
Product Link https://www.ambeed.com/products/808755-53-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/808755-53-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart