AMBH58064562 Ambeed, Inc.

(2S,4S)-tert-Butyl 4-amino-2-methylpyrrolidine-1-carboxylate

  • MDL number MFCD27945257
  • Cas Number : 152673-32-0
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/152673-32-0.html

Safety Information

MSDS https://www.ambeed.com/products/152673-32-0.html
Availability details for
Quantity
  • Availability
  • Add to Cart