AMBH5806456B Ambeed, Inc.

(1R,2R)-rel-Ethyl 2-phenylcyclopropanecarboxylate

  • MDL number MFCD00019221
  • Cas Number : 946-39-4
  • Molecular Weight : 190.2384

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/946-39-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/946-39-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart