AMBH58064574 Ambeed, Inc.

Ethyl 5,7-dichloroimidazo[1,2-c]pyrimidine-2-carboxylate

  • MDL number MFCD18837678
  • Cas Number : 1289151-91-2
  • Molecular Weight : 260.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/1289151-91-2.html

Safety Information

MSDS https://www.ambeed.com/products/1289151-91-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart