AMBH58064576 Ambeed, Inc.

7-(Diethylamino)-4-(hydroxymethyl)-2H-chromen-2-one

  • MDL number N/A
  • Cas Number : 54711-38-5
  • Molecular Weight : 247.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/54711-38-5.html

Safety Information

MSDS https://www.ambeed.com/products/54711-38-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart