AMBH58064577 Ambeed, Inc.

2-Methyl-1-(2-nitrophenyl)-1H-imidazole

  • MDL number MFCD13689003
  • Cas Number : 26286-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/26286-51-1.html

Safety Information

MSDS https://www.ambeed.com/products/26286-51-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart