AMBH5806457C Ambeed, Inc.

Calcium butyrate

  • MDL number N/A
  • Cas Number : 5743-36-2
  • Molecular Weight : 214.2724

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/5743-36-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/5743-36-2.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart