AMBH58064580 Ambeed, Inc.

trans-1,2-Dithiane-4,5-diol

  • MDL number MFCD00006661
  • Cas Number : 14193-38-5
  • Molecular Weight : 152.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/14193-38-5.html

Safety Information

MSDS https://www.ambeed.com/products/14193-38-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart