AMBH58064584 Ambeed, Inc.

(5-(Methylsulfonyl)pyridin-3-yl)methanamine

  • MDL number MFCD18256699
  • Cas Number : 1211584-23-4
  • Molecular Weight : 186.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1211584-23-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1211584-23-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart