AMBH5806458A Ambeed, Inc.

2-Bromo-1,3-difluoro-5-iodobenzene

  • MDL number MFCD06797752
  • Cas Number : 155906-10-8
  • Molecular Weight : 318.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/155906-10-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/155906-10-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart