AMBH5806458D Ambeed, Inc.

SAR7334

  • MDL number MFCD27992059
  • Cas Number : 1333210-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin China
Product Link https://www.ambeed.com/products/1333210-07-3.html

Safety Information

MSDS https://www.ambeed.com/products/1333210-07-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart