AMBH5806458F Ambeed, Inc.

BX471

  • MDL number MFCD09859709
  • Cas Number : 217645-70-0
  • Molecular Weight : 434.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/217645-70-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/217645-70-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart