AMBH58064592 Ambeed, Inc.

(S)-1-Boc-3-Aminomethylpyrrolidine hydrochloride

  • MDL number MFCD11101250
  • Cas Number : 916214-30-7
  • Molecular Weight : 236.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/916214-30-7.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/916214-30-7.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart