AMBH58064596 Ambeed, Inc.

4-Methoxycyclohex-3-enol

  • MDL number MFCD11656596
  • Cas Number : 69125-55-9
  • Molecular Weight : 128.1690

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/69125-55-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/69125-55-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart