AMBH5806459C Ambeed, Inc.

1-(Bromomethyl)-3-chloro-2-nitrobenzene

  • MDL number MFCD17168787
  • Cas Number : 56433-00-2
  • Molecular Weight : 250.4771

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form Solid or liquid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/56433-00-2.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/56433-00-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart