AMBH5806459D Ambeed, Inc.

(S)-1-(5-Methylfuran-2-yl)propan-1-amine

  • MDL number MFCD11973975
  • Cas Number : 473732-95-5
  • Molecular Weight : 139.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/473732-95-5.html

Safety Information

MSDS https://www.ambeed.com/products/473732-95-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart