AMBH5806459E Ambeed, Inc.

2,4-Dimethylbenzenesulfonic acid monohydrate

  • MDL number MFCD00673743
  • Cas Number : 58811-92-0
  • Molecular Weight : 186.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/58811-92-0.html

Safety Information

signalword Warning
pcodes P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313
MSDS https://www.ambeed.com/products/58811-92-0.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart