AMBH5806459F Ambeed, Inc.

(4-Bromo-1-methyl-1H-pyrazol-3-yl)methanol

  • MDL number MFCD06202888
  • Cas Number : 915707-65-2
  • Molecular Weight : 191.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 94
bp 308.4�t 760 mmHg
Iupac Name (4-bromo-1-methyl-1H-pyrazol-3-yl)methanol
Inchi Code 1S/C5H7BrN2O/c1-8-2-4(6)5(3-9)7-8/h2,9H,3H2,1H3
Inchi Key JFGLJTFTVBZOCB-UHFFFAOYSA-N

Description

Melting Point 94
Purity 98%
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 308.4�t 760 mmHg
Product Link https://www.ambeed.com/products/915707-65-2.html

Safety Information

signalword Warning
pcodes P261-P264-P270-P271-P280-P301+P312+P330-P302+P352+P312-P304+P340+P312-P305+P351+P338-P332+P313-P337+P313-P362-P363-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/915707-65-2.html
pictograms GHS07
hstatements H302-H315-H319-H335-H312-H332
Availability details for
Quantity
  • Availability
  • Add to Cart