AMBH580645A5 Ambeed, Inc.

Methyl 2-fluoro-3,5-dimethoxybenzoate

  • MDL number MFCD18382551
  • Cas Number : 651734-58-6
  • Molecular Weight : 214.1904

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/651734-58-6.html

Safety Information

signalword Warning
pcodes P264-P270-P301+P312-P330
MSDS https://www.ambeed.com/products/651734-58-6.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart