AMBH580645B1 Ambeed, Inc.

6-Fluoro-2,3-dihydrobenzofuran-3-amine

  • MDL number MFCD10696192
  • Cas Number : 944904-32-9
  • Molecular Weight : 153.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/944904-32-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/944904-32-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart