AMBH580645B4 Ambeed, Inc.

Ethyl 2,3,4,6-Tetra-O-acetyl-�D-thioglucopyranoside

  • MDL number MFCD00797608
  • Cas Number : 41670-79-5
  • Molecular Weight : 392.4200

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/41670-79-5.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/41670-79-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart