AMBH580645B8 Ambeed, Inc.

1-Bromo-8-chloronaphthalene

  • MDL number MFCD17012231
  • Cas Number : 20816-79-9
  • Molecular Weight : 241.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/20816-79-9.html

Safety Information

MSDS https://www.ambeed.com/products/20816-79-9.html
Availability details for
Quantity
  • Availability
  • Add to Cart