AMBH580645BC Ambeed, Inc.

NOD-IN-1

  • MDL number MFCD30496705
  • Cas Number : 132819-92-2
  • Molecular Weight : 343.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/132819-92-2.html

Safety Information

MSDS https://www.ambeed.com/products/132819-92-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart