AMBH580645BF Ambeed, Inc.

(R)-1-Benzyl 4-tert-butyl 2-(hydroxymethyl)piperazine-1,4-dicarboxylate

  • MDL number MFCD09261339
  • Cas Number : 1217813-68-7
  • Molecular Weight : 350.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1217813-68-7.html

Safety Information

MSDS https://www.ambeed.com/products/1217813-68-7.html
Availability details for
Quantity
  • Availability
  • Add to Cart