AMBH580645CA Ambeed, Inc.

2-Fluoro-1H-imidazole hydrochloride

  • MDL number MFCD24369564
  • Cas Number : 42309-92-2
  • Molecular Weight : 122.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/42309-92-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/42309-92-2.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart