AMBH580645D2 Ambeed, Inc.

3-(Carboxymethyl)-5-chloro-1H-indole-2-carboxylic acid

  • MDL number MFCD00897388
  • Cas Number : 120716-95-2
  • Molecular Weight : 253.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Product Link https://www.ambeed.com/products/120716-95-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/120716-95-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart